Order Entry
United States
Orders LinkContactUsLinkComponent
 

Silver Nitrate

Silver Nitrate, Solution, Laboratory, 100 mL
  470302-396
 :  SS0161-100ML
Silver Nitrate
  470302-396
 :  
Silver Nitrate
Silver Nitrate

Silver Nitrate, Solution, Laboratory, 100 mL

 :  
 
$330.00
   
Each (100ml)
 
   
 

CAS Number: 7761-88-8
Formula: AgNO3
Density: 4.35 g/mL
Freezing Point: 212 °C
Solubility: Water, Hot Alcohol
Shelf Life: 24 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

 : This product is a corrosive, toxic irritant.

 

 

  • Description:
    Silver Nitrate, Solution
  • Size:
    100 mL
  • Chemical/physical specifications:
    1.0 M
  • Grade:
    Laboratory

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.