Order Entry
United States
Orders LinkContactUsLinkComponent
Cooling Containers

Cooling Containers

Pan, 9 L, Red
Catalog # 490007-650
Supplier:   Heathrow Scientific
Supplier #:  HS28729R
Cooling Containers
Catalog # 490007-650
Cooling Containers
Cooling Containers

Pan, 9 L, Red

Price
$246.30
New Customer Promotional Price
Log in for availability
Each Case of 6
Product Details & Documents

Durable containers for chilling temperature-sensitive samples.

  • Made from polyurethane with metal inserts for stability
  • Close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob
  • Tapered design for easy stacking
  • Precise pour spout and large handles
  • The lid and bottom of the bucket/pan are keyed to fit together
  • For low-temperature use to −80 °C

Suitable for low temperature use. Ideal for ice, ice slurries and CO₂ pellets.

Specifications
  • Capacity:
    9 L
  • Color:
    Red
  • Description:
    Pan