Order Entry
United States
Orders LinkContactUsLinkComponent
 

Advansta Buffer Pouches

Buffer PBS Pouches
Catalog # 490005-044
Supplier:   Advansta
 :  R-01038-020
Advansta Buffer Pouches
Catalog # 490005-044
Supplier:  Advansta
CAS Number:  
Advansta Buffer Pouches
Advansta Buffer Pouches

Buffer PBS Pouches

CAS Number:  
Price
$140.96
Log in for availability
Pack of 20
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Convenient, pure, fast buffer preparation

  • Quality- Molecular biology-grade buffers of highest purity
  • Convenient- Pre-measured powder in sealed pouches
  • Fast- Dissolve and use right away
  • Standardized- Guaranteed reproducible results for your electrophoresis gels and Western blots

Advansta's Avant buffer pouches are perfect for standardizing your electrophoresis and Western blotting applications. Dissolve a pouch of pre-messured molecular biology-grade chemicals in 500 mL of deionized water to obtain a ready-to-use 1X buffer without any further preparation. Save precious time in the laboratory and generate reproducible results with Avant buffers.

Specifications

Specifications

  • Description:
    Buffer PBS Pouches

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.