pH Gradient Buffers
CX-1 pH gradient buffer B, 125 ml, 10.2
Catalog # 10053-804
Supplier: Thermo Scientific
Supplier #: 083275
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Deliver robust, reproducible gradients with the Thermo Scientific pH buffer platform, which is applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.
- Proprietary buffer formulations enable fast, robust and reproducible pH gradients that are simple to optimise and easily automated
- Ready to use with existing LC columns and systems, without the need for time-consuming mobile phase adjustments
- Applicable to the majority of MAbs
The pH gradient platform saves time in method development and facilitates method transfer to QA/QC for a wide range of MAb charge variants through a generic LC-based approach to MAb characterization.
All that is required to generate a pH gradient is a 1:10 dilution of the pH buffer concentrates. A linear pH gradient from pH 5.6 to 10.2 can simply be delivered by running a pump gradient from 100% eluent A to 100% eluent B.
Documents
Specifications
Specifications
- Description:CX-1 pH gradient buffer B
- Size:125 ml
- pH:10.2



