Orders LinkContactUsLinkComponent
pH Gradient Buffers
pH Gradient Buffers
CX-1 pH gradient buffer A, 125 ml, 5.6
  10053-800
Supplier #:  083273
pH Gradient Buffers
  10053-800
 :  
pH Gradient Buffers
CX-1 pH gradient buffer A, 125 ml, 5.6
 :  
$276.78
Each
Deliver robust, reproducible gradients with the Thermo Scientific pH buffer platform, which is applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.
  • Proprietary buffer formulations enable fast, robust and reproducible pH gradients that are simple to optimise and easily automated
  • Ready to use with existing LC columns and systems, without the need for time-consuming mobile phase adjustments
  • Applicable to the majority of MAbs
The pH gradient platform saves time in method development and facilitates method transfer to QA/QC for a wide range of MAb charge variants through a generic LC-based approach to MAb characterization.
All that is required to generate a pH gradient is a 1:10 dilution of the pH buffer concentrates. A linear pH gradient from pH 5.6 to 10.2 can simply be delivered by running a pump gradient from 100% eluent A to 100% eluent B.
  • Description:
    CX-1 pH gradient buffer A
  • Size:
    125 ml
  • pH:
    5.6