StatSpin® Microhematocrit Tubes, Iris Sample Processing
SafeCrit plastic capillary tube, Sodium Heparin anticoagulant, 40 mm, 20 µl
Supplier: Beckman Coulter
: HP8H-10
StatSpin® Microhematocrit Tubes, Iris Sample Processing
Catalog # 87002-149
Supplier: Beckman Coulter
Jump to:
Product Details & Documents
These microhematocrit tubes use capillary action to draw samples quickly.
- Capillary Action Draws Samples Quickly
- Material: Non-Breakable Plastic for Safety
- Heparin Tubes for Finger-Stick Samples
- Untreated Tubes for Anti-Coagulated Venous Samples
- Available Lengths: 75 mm & 40 mm
Tubes are available with heparin for finger-stick samples or untreated for anti-coagulated venous samples
SafeCrit® hematocrit tubes are made of non-breakable, 100% plastic to eliminate the potential safety hazards of broken glass.
The 40 mm tubes are made for use with the MP multipurpose centrifuge (87002-248 series) and the CritSpin™ microhematocrit centrifuge (87002-206 series). The 75 mm tubes are made for use in conventional centrifuges that support the full length of the tube.
Specifications
- Volume:20 µl
- Description:SafeCrit plastic capillary tube, Sodium Heparin anticoagulant
- Length:40 mm



