Order Entry
United States
Orders LinkContactUsLinkComponent
 

StatSpin® Microhematocrit Tubes, Iris Sample Processing

SafeCrit plastic capillary tube, no anticoagulant, 75 mm, 40 µl
Catalog # 87002-147
Supplier:   Hemocue
 :  HP4U-10
StatSpin® Microhematocrit Tubes, Iris Sample Processing
Catalog # 87002-147
Supplier:  Hemocue
StatSpin® Microhematocrit Tubes, Iris Sample Processing
StatSpin® Microhematocrit Tubes, Iris Sample Processing

SafeCrit plastic capillary tube, no anticoagulant, 75 mm, 40 µl

Price
$34.81
Log in for availability
Pack of 100 Case of 1000
Product Details & Documents

These microhematocrit tubes use capillary action to draw samples quickly.

  • Capillary Action Draws Samples Quickly
  • Material: Non-Breakable Plastic for Safety
  • Heparin Tubes for Finger-Stick Samples
  • Untreated Tubes for Anti-Coagulated Venous Samples
  • Available Lengths: 75 mm & 40 mm

Tubes are available with heparin for finger-stick samples or untreated for anti-coagulated venous samples

SafeCrit® hematocrit tubes are made of non-breakable, 100% plastic to eliminate the potential safety hazards of broken glass.

The 40 mm tubes are made for use with the MP multipurpose centrifuge (87002-248 series) and the CritSpin™ microhematocrit centrifuge (87002-206 series). The 75 mm tubes are made for use in conventional centrifuges that support the full length of the tube.

Specifications
  • Volume:
    40 µl
  • Description:
    SafeCrit plastic capillary tube, no anticoagulant
  • Length:
    75 mm