COD Standard Solutions, CRM traceable to SRM from NIST
COD standard solution, 100 ml, 8000 mg/L, Plastic Bottle
Supplier: MilliporeSigma
Supplier #: 1250340100
COD Standard Solutions, CRM traceable to SRM from NIST
Catalog # EM1.25034.0100
Supplier: MilliporeSigma
CAS Number:
Jump to:
Product Details & Documents
- Diluted standard solutions that are ready-to-use for accurate measurements
- Traceable to SRM from NIST
- Full certificates of analysis with comprehensive quality documentation
- Information on concentration and expanded measurement uncertainty
- Save time and minimize errors
Specifications
Specifications
- Concentration:8000 mg/L
- Pack type:Plastic Bottle
- Description:COD standard solution
- Size:100 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



