Order Entry
United States
Orders LinkContactUsLinkComponent
COD Standard Solutions, CRM traceable to SRM from NIST

COD Standard Solutions, CRM traceable to SRM from NIST

COD standard solution, 100 ml, 2000 mg/L, Plastic Bottle
Catalog # EM1.25033.0100
Supplier:   MilliporeSigma
Supplier #:  1250330100
COD Standard Solutions, CRM traceable to SRM from NIST
Catalog # EM1.25033.0100
Supplier:  MilliporeSigma
CAS Number:  
COD Standard Solutions, CRM traceable to SRM from NIST

COD standard solution, 100 ml, 2000 mg/L, Plastic Bottle

CAS Number:  
Price
$80.19
Log in for availability
Each (100ml) Case of 42 (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Specifications

Specifications

  • Concentration:
    2000 mg/L
  • Pack type:
    Plastic Bottle
  • Description:
    COD standard solution
  • Size:
    100 ml
  • Cat. No.:
    EM1.25033.0100

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.