Order Entry
United States
Orders LinkContactUsLinkComponent
Cell-Chex Controls, Streck

Cell-Chex Controls, Streck

Level 1-UC, Level 1-CC, and Level 2, 3 each×2.0 ml
Catalog # 10029-560
Supplier:   Streck Laboratories
Supplier #:  212431
Cell-Chex Controls, Streck
Catalog # 10029-560
CAS Number:  
Cell-Chex Controls, Streck
Cell-Chex Controls, Streck

Level 1-UC, Level 1-CC, and Level 2, 3 each×2.0 ml

CAS Number:  
Price
$200.72
Log in for availability
Each (6ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cell-Chex controls offer spinal and body fluid procedural control for red blood cell and white blood cell counts, crystal identification and white blood cell differentiation

Level 1-UC contains urate crystals, and Level 1-CC contains CPPD crystals to aid in identification of crystals in synovial fluid. The white blood cells can be differentiated when stained in the same manner as a patient sample. The two-level control is assayed for manual cell counting, five-part white blood cell differential and two-part polymorphonuclear and mononuclear differential

Ordering information: Cell-Chex controls maintain a closed-vial stability for 6 months and an open-vial stability for 30 days when stored at 2 to 10°C (36 to 50°F).

Specifications

Specifications

  • Description:
    Level 1-UC, Level 1-CC, and Level 2
  • Size:
    3 each×2.0 ml

Specifications