Oakton pH 5+ and 6+ Handheld pH Meters, Cole-Parmer
Catalog # 101076-426
Supplier: OAKTON INSTRUMENTS MS
Specifications
- Temperature Range:−10...+110 °C
- Description:Oakton pH 5+ Handheld Meter Kit w/ Case, Solutions, pH/ATC Probe
- Model:pH 5+
- Dimensions:3 3/8 × 1 11/16"
- Display:LCD
- Power supply:4 AAA batteries (included)
- Temperature Compensation:Automatic
- pH Accuracy:±0.01 pH
- pH Calibration:Up to 5 points
- pH Range:0 - 14
- pH Resolution:0.01 or 0.1
- Cat. No.:101076-426
- For use with:Oakton pH electrodes with BNC connector
- Supplier no.:WD-35613-54
Specifications
About this item
High ±0.01 pH accuracy at an economical price.
- Convenient, compact design
- Up to five-point push-button calibration with autobuffer recognition
- Displays pH and temperature with the press of a button
- Automatic temperature compensation (ATC) ensures accuracy from 0 to 80°C
Meters include a protective rubber boot that also acts as a meter stand.
Ordering information: Includes All-in-one pH electrode, ATC probe, and batteries. Kits add: pH buffer solutions, sample bottles, rinse bottle, protective rubber boot, and carrying case.
Frequently Bought Together