Human Recombinant M-Csf
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
- Consistent Sizing options
- Datasheet Included
- High Purity, Low Endotoxin levels
Recombinant Human Macrophage Colony-Stimulating Factor is produced by our Mammalian expression system and the target gene encoding Glu33-Arg255 is expressed with a 6His tag at the C-terminus.
: Accessories not included.
: Shipped Via FedEx
: Shipped at ambient temperature via FedEx envelope.
: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
- Size:10 μg
- Cat. No.:102861-974
- Enzyme type:Recombinant
- Species:Human
- Enzyme Name:M-Csf