Order Entry
United States
Orders LinkContactUsLinkComponent
Fluid A, USP

Fluid A, USP

Fluid A, USP, —, 200 ml, Serum bottle, 10
Catalog # 89407-529
Fluid A, USP
Catalog # 89407-529
Supplier:  
CAS Number:  
Fluid A, USP
Fluid A, USP

Fluid A, USP, —, 200 ml, Serum bottle, 10

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BOTTLE W/SEP F/STERILTY PK10 Catalog #90004-139

Product 90004-139 is a replacement for 89407-529 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

A rinsing and diluting fluid for use in USP testing protocols.

A rinsing and diluting fluid for use in USP testing protocols. Contains peptone 0.1%.

Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center. Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.

Ordering information: Order by the pack of 10

Specifications

Specifications

  • Pack type:
    Serum bottle
  • Description:
    Fluid A, USP
  • Size:
  • Minimum Order Quantity:
    10
  • Volume:
    200 ml

Specifications