Order Entry
United States
Orders LinkContactUsLinkComponent
 

MColortest™ pH-indicator solution pH 4.0 - 10.0 Universal Indicator with colour Card

100 ml, pH 4-4,5-5-5,5-6-6,5-7-7,5-8-8,5-9-9,5-10, Plastic bottle
Catalog # EM1.09175.0100
Supplier:   MilliporeSigma
 :  1091750100
MColortest™ pH-indicator solution pH 4.0 - 10.0 Universal Indicator with colour Card
Catalog # EM1.09175.0100
Supplier:  MilliporeSigma
CAS Number:  
MColortest™ pH-indicator solution pH 4.0 - 10.0 Universal Indicator with colour Card

100 ml, pH 4-4,5-5-5,5-6-6,5-7-7,5-8-8,5-9-9,5-10, Plastic bottle

CAS Number:  
Price
$154.59
Log in for availability
Each (100ml) Case of 35 (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.

  • Results in approx. 1 Minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable

 : Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 ml
  • Range:
    pH 4-4,5-5-5,5-6-6,5-7-7,5-8-8,5-9-9,5-10

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.