Order Entry
United States
Orders LinkContactUsLinkComponent
 

3-Ethoxypropionic acid ≥97%

10 g
Catalog # 102838-742
Marketsource
Supplier:   Matrix Scientific
 :  083039-10G
3-Ethoxypropionic acid ≥97%
Catalog # 102838-742
Supplier:  Matrix Scientific
Marketsource
CAS Number:  4324-38-3
3-Ethoxypropionic acid ≥97%

10 g

CAS Number:  4324-38-3
Price
Inquire For Price
Log in for availability
Each (10g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Hazard: Irritant

  • CAS Number:4324-38-3
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00058997
Specifications

Specifications

  • Size:
    10 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.