Boron trifluoride diethyl ether complex ≥98%
Product Details & Documents
CAS Number: 109-63-7 Molecular Formula: C4H10BF3O Molecular Weight: 141.93 Form: Liquid Color: Colorless
Product Categories: Analytical Science, NMR Spectrometry, Bioscience, Sugars, Synthetic Organic Chemistry, Carbonyl Olefination, Functional Group Transformation and Introducing Reaction, Multicomponent Reaction (MCR)
- CAS Number:109-63-7
- Formula:C₄H₁₀BF₃O
- MDL Number:MFCD00013194
- Molecular Weight:141.93 g/mol
- Merck Index:13,01340
- Boiling Pt:126 °C (1013 hPa)
- Melting Pt:–60 °C
- Density:1.125 g/cm³ (25 °C)
- Flash Pt:59 °C
- Storage Temperature:Ambient
- EINECS:203-689-8
- UN:2604
- ADR:8,I
Specifications
Specification Test Results
- Purity(Gravimetric Method):98.0 to 102.0 %
- Specific gravity (20/20):1.1250 to 1.1320
- Refractive index n20/D:1.3430 to 1.3470
Specifications
- Size:25 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



