Order Entry
United States
Orders LinkContactUsLinkComponent
Crystal violet

Crystal violet

25 g
Catalog # TCC0428-25G
Corrosive
Health
Irritant
Environment
Supplier:   TCI America
Supplier #:  C0428-25G
Crystal violet
Catalog # TCC0428-25G
Supplier:  TCI America
CAS Number:  548-62-9
Crystal violet

25 g

CAS Number:  548-62-9
Price
$45.00
Log in for availability
Each (25g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 548-62-9
MDL Number: MFCD00011750
Molecular Formula: C25H30ClN3
Molecular Weight: 407.99
Form: Crystal
Color: Yellow Green
Melting point (°C): 211

Other Product Names / Synonyms: Basic Violet 3, Hexamethylpararosaniline Chloride

Synonyms:  Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
  • CAS Number:548-62-9
  • Formula:C₂₅H₃₀ClN₃
  • MDL Number:MFCD00011750
  • Molecular Weight:407.99 g/mol
  • Merck Index:13,04406
  • Melting Pt:211 °C
  • Density:1.19 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:208-953-6
  • UN:3077
  • ADR:9,III

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.