Order Entry
United States
Orders LinkContactUsLinkComponent
Gentamicin sulfate

Gentamicin sulfate

5 g
Catalog # TCG0383-5G
Supplier:   TCI America
Supplier #:  G0383-5G
Gentamicin sulfate
Catalog # TCG0383-5G
Supplier:  TCI America
CAS Number:  1405-41-0
Gentamicin sulfate

5 g

CAS Number:  1405-41-0
Price
$211.00
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Gentamicin, an aminoglycoside antibiotic, is isolated from Micromonospora purpurea, a Gram-positive bacteria

  • Pale yellow crystals

Gentamicin has activity against mostly Gram-negative bacteria including Pseudomonas, Proteus, Serratia and the Gram-positive Staphylococcus. Gentamicin is widely used because of its potent bactericidal activities and less bacterial resistance and is well known to cause nephrotoxicity and hypacusis.

It consists of a mixture of three isomers, gentamicin C1, gentamicin C1a and gentamicin C2. Gentamicin interacts with the 30S subunit of prokaryotic ribosomes, and then it inhibits bacterial growth by inhibiting protein synthesis.

Synonyms:  Gentamicin sulfate salt, NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS Number:1405-41-0
  • MDL Number:MFCD00270181
  • Storage Temperature:Refrigerator

Documents

Specifications

Specifications

  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.