Product Details & Documents
Accurate pH Testing
- 100 Tests
- Spot Testing of Your Soil
Tests are made with a wide range indicator and give the pH of soil to ±0.5 pH units. Includes 50 tests, chemical indicator, porcelain plates, color comparator chart, complete Instructions, pocket-size case and a soil test handbook.
Documents
ghs_5024.pdf
SDS
Specifications
Specifications
- Description:Soil Acidity Test Kit
- Range:4 - 8 pH