Orders LinkContactUsLinkComponent
Master Forensics Kit
Master Forensics Kit
Catalog # 470200-748
Master Forensics Kit
Catalog # 470200-748
Supplier:  
Master Forensics Kit
Master Forensics Kit
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
REFLL MASTER FORENSICS 36-5897 24 STUD. Catalog #470015-370
Product 470015-370 is a replacement for 470200-748 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Enough materials for an entire unit on crime scene investigation.
  • Grades 7-14
  • Combination of 5 kits
Apply the disciplines of biology, chemistry, and physics with the materials and procedures introduced in this activity. Several techniques used in forensics are covered and include the analysis of fingerprints, soil, hair, documents, and drugs and poisoning.
It comes with enough materials for six setups and an instruction manual.
This Master Forensics Kit includes truncated versions of our Fingerprint Analysis, Soil and Mineral Analysis, Hair Analysis, Document Analysis, and Analysis of Drugs and Poisons kits, and culminates with an extensive crime scene activity that reinforces learned techniques. Following training, your CSI investigators engage in the ultimate hands on assessment activity. They watch the included video setting the scene for the crime and collect and analyze evidence to solve one of four possible case scenarios. Includes all materials for training activities, a faculty cast list, and materials for preparing the crime scene evidence box. Complete with an extensive instructor’s manual, flash drives, reproducible student instructions, and reproducible worksheets. Allow the equivalent of 12-14 one-hour lab periods. You will need to supply Microscopes, Chemical Splash Goggles, UV light, Oven or Heat Lamp, balance, (36) large test tubes, (6) 10 ml graduated cylinders, (6) evaporating dishes, gloves and lab aprons. Designed for 24 students working in groups of four.
Documents