Orders LinkContactUsLinkComponent
Ward's® Fingerprint Classification Chart
Ward's® Fingerprint Classification Chart
Catalog # 470199-716
Supplier:   Avantor
Ward's® Fingerprint Classification Chart
Catalog # 470199-716
Supplier:  Avantor
Ward's® Fingerprint Classification Chart
Ward's® Fingerprint Classification Chart
Clearance
$29.99
Log in for availability
Each
Product Details & Documents
Learn How Forensic Scientists Classify Types
  • Size: 21 x 34"
Display detailed images and definitions of various fingerprint patterns on your walls for your students to study. The poster shows loops, arches, and whorls, as well as specific friction ridge characteristics such as bifurcation, enclosure, ridge endings, and more. Size: 21" x 34".