Tris-Borate-EDTA Solution
Tris-Borate-EDTA Running Buffer, Laboratory, Makes 6 L
Supplier: Ward's Science
Supplier #: TT0283-102G
Jump to:
Product Details & Documents
Tris-Borate-EDTA Solution is premixed.
- Powder form
- For use with Borate gels only
- Premixed
It is in powder form.
The solution is for use with borate gels only.
Certifications: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Specifications
Specifications
- Description:Tris-Borate-EDTA Running Buffer
- Size:Makes 6 L
- Chemical/physical specifications:1X Solution
- Grade:Laboratory



