Orders LinkContactUsLinkComponent
Modular Display Cases
Modular Display Cases
Catalog # 470117-074
Modular Display Cases
Catalog # 470117-074
Supplier:  
Modular Display Cases
Discontinued
Product Details & Documents
The floor model case with base provides 14" depth display storage within the glass case. Ships with one glass shelf. Stack a second case on top of the Case/Base combination for extra display space. Base is 10" deep for stability.
  • Specify color when ordering. Overall Dimensions of Casewith Base: 36"L×14"D×40"H. Add-on Case only: 36"L×14"D×30"H
Cases are available with white laminate, mirror or fabric back. Case finishes available in Honey Maple, Danish Walnut, and Cordovan.