Orders LinkContactUsLinkComponent
Pipe Stem Triangles
Pipe Stem Triangles
Pipe Stem Triangle, side length 6 cm (2.5")
Catalog # 470189-156
Supplier #:  1350627020000
Pipe Stem Triangles
Catalog # 470189-156
Pipe Stem Triangles
Pipe Stem Triangles
Pipe Stem Triangle, side length 6 cm (2.5")
Price
$4.24
Log in for availability
Each Pack of 12
Product Details & Documents
Triangles made from galvanized iron wire, with porcelain pipe stemming to reduce wire sagging and oxidation.
  • Heavy gauge wire
  • Galvanized steel
Provides a patform to hold items over a Bunsen burner.
Specifications
  • Description:
    Pipe Stem Triangle, side length 6 cm (2.5")