Pipe Stem Triangles
Pipe Stem Triangle, side length 4 cm (1.5")
470189-154
: CPT015
Triangles made from galvanized iron wire, with porcelain pipe stemming to reduce wire sagging and oxidation.
- Heavy gauge wire
- Galvanized steel
Provides a patform to hold items over a Bunsen burner.
- Description:Pipe Stem Triangle, side length 4 cm (1.5")