Slide Grid
Slide Grid tool
Catalog # 470092-736
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
GRAPH, BLACK GRIDLINES, (SLIDE A) Catalog #470320-874
Product 470320-874 is similar to 470092-736 but may have a different manufacturer and/or packaging.
Product Details & Documents
Microscopy tool to estimate size and counts.
- Microscopy study and research tool
- Improve your counting estimates
Use this slide grid as a tool to count or measure cells or organisms or to estimate the size of microscopic organisms. The grid is 1 cm square with mm divisions.
Specifications
- Description:Slide Grid tool