Orders LinkContactUsLinkComponent
Live Restriction Enzymes
Live Restriction Enzymes
Live Hind III, 50 μl
Catalog # 470176-810
Supplier:   Avantor
Live Restriction Enzymes
Catalog # 470176-810
Supplier:  Avantor
CAS Number:  
Live Restriction Enzymes
Live Hind III, 50 μl
CAS Number:  
Price
$24.99
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
These restriction enzymes are used to recognize specific sequences in double strand DNA. BamH I recognizes 5-G/GATCC-3, Ecor I recognizes 5-G/AATTC-3, and Hind III recognizes 5-A/AGCTT-3. The activity level of all our restriction enzymes is 10 units per 1 µl.
All are shipped next day on ice. As they are perishable, a ship date must be specified.
Specifications
Specifications
  • Description:
    Live Hind III
  • Size:
    50 μl