Orders LinkContactUsLinkComponent
Disposable Specimen Containers
Disposable Specimen Containers
Specimen Container, 1.81 L, Disposable
  470173-878
Supplier #:  453625
Disposable Specimen Containers
  470173-878
 :  
Disposable Specimen Containers
Specimen Container, 1.81 L, Disposable
Keep specimens organized and out of your way with these handy storage units.
Featuring high-density polyethylene construction and tight-fitting lids, these translucent containers are resistant to freezing and boiling, and are sturdy enough to be used multiple times.
  • Description:
    Specimen Container, 1.81 L, Disposable