Order Entry
United States
Orders LinkContactUsLinkComponent
 

Buffer Capsules

Buffer Capsules, Laboratory, Vial/10
  470158-208
 :  270-PH09
Buffer Capsules
  470158-208
 :  
Buffer Capsules
Buffer Capsules

Buffer Capsules, Laboratory, Vial/10

 :  
 
$13.30
   
Each Case of 50
 
   
 

Shelf Life (months): 36
Storage: Green

 : Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

 

 

  • Description:
    Buffer Capsules
  • Size:
    Vial/10
  • Chemical/physical specifications:
    pH 9.00
  • Grade:
    Laboratory

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.