Order Entry
United States
Orders LinkContactUsLinkComponent
Buffer Capsules

Buffer Capsules

Buffer Capsules, Laboratory, Vial/10
Catalog # 470158-200
Supplier:   Micro Essential
Supplier #:  270-PH11
Buffer Capsules
Catalog # 470158-200
Supplier:  Micro Essential
CAS Number:  
Buffer Capsules
Buffer Capsules

Buffer Capsules, Laboratory, Vial/10

CAS Number:  
Price
$13.30
Log in for availability
Each Case of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Shelf Life (months): 36
Storage: Green

Caution: Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Buffer Capsules
  • Size:
    Vial/10
  • Chemical/physical specifications:
    pH 11.00
  • Grade:
    Laboratory

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.