Order Entry
United States
Orders LinkContactUsLinkComponent
Plastic Forceps

Plastic Forceps

Plastic Forceps, Plastic, 140 mm (5.5"), Straight
Catalog # 470156-704
Supplier:   C & A Scientific
Supplier #:  97-3906
Plastic Forceps
Catalog # 470156-704
Supplier:  C & A Scientific
Plastic Forceps
Plastic Forceps

Plastic Forceps, Plastic, 140 mm (5.5"), Straight

Clearance
$1.25
Sale

Valid until 31/12/2026

Log in for availability
Pack of 6 Case of 600
Product Details & Documents

Rust-free performance.

  • Plastic construction for durability
  • Textured tip for reduced slippage
  • Textured surface for easy control
  • Aligned with GLOBE protocols

These plastic forceps feature serrated tips with a wide handle.

Specifications
  • Description:
    Plastic Forceps
  • Length:
    140 mm (5.5")
  • Material:
    Plastic
  • Tip style:
    Straight
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.