Order Entry
United States
Orders LinkContactUsLinkComponent
 

Skinned Cats

  470154-134
Skinned Cats
  470154-134
 :  
Skinned Cats
Skinned Cats
 

Standard formalin preserved cats, packed in vaccum-sealed bags. One cat per bag.

  • Vertebrate anatomy
  • Mammalian anatomy
  • High quality preservation

Cats, Felis species are expertly preserved and presented in an extended position for anatomical correctness and ease of study. Our latex injected cats are injected with elastic latex with bright, contrasting colors, to ensure ease of dissection. Plastic storage bags and waterproof ID tags are provided free with each cat. Gender selection can be made upon request but depends on availability. Call for details.

 : Cats are sourced from pounds and shelters. All shelters are required to retain animals for a specified time to maximize the possibility of adoption. Animals that are not adopted are euthanized at the shelters and preserved for dissection under the guidance of USDA regulations. These animals are "recycled" for education; otherwise they would have been incinerated or placed in a landfill after euthanasia.

Accessories

 

  •  
    Description
    Preservative
    Size
     
     
  • 470001-050
    Skinned Rabbit
    Formalin preserved
    14 to 19"
       
    Each - $79.95

 

  •  
    Description
    Preservative
    Size
     
     
  • 470001-040
    Rabbit
    Formalin preserved
    14 to 19"
       
    Each - $69.95

 

  •  
    Description
    Preservative
    Size
     
     
  • 470001-044
    Skinned Rabbit
    Formalin preserved
    14 to 19"
       
    Each - $74.95

 

  •  
    Description
    Preservative
    Size
     
     
  • 470001-034
    Skinned Rabbit
    Formalin preserved
    14 to 19"
       
    Each - $69.95

 

  •  
    Description
    Preservative
    Size
     
     
  • 470001-030
    Rabbit
    Formalin preserved
    14 to 19"
       
    Each - $59.95

 

  •  
    Description
    Preservative
    Size
     
     
  • 470001-026
    Skinned Rabbit
    Formalin preserved
    14 to 19"
       
    Each - $64.95

 

  •  
    Description
    Preservative
    Size
     
     
  • 470001-022
    Rabbit
    Formalin preserved
    14 to 19"
       
    Each - $64.95

 

  •  
    Description
     
     
  • 470220-350
    Bookstore Dissection Kit
       
    Each - $42.40

 

  •  
    Description
    Grade
    Size
     
     
  • 470233-546
    WARDSafe™
    Laboratory
    19 L
       
    Each (19l) - $218.90