pH Indicator Papers
pH Indicator Papers, Litmus Blue, Laboratory, Strips, Strips, Laboratory
Catalog # 470150-838
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PAPER LITMUS BLUE VL Catalog #470123-117
Product 470123-117 is a replacement for 470150-838 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
pH Test Papers in your Choice of Two Indicator Colors.
- Easy to read
- Vial of 100
Litmus red paper changes to blue at pH 8.1. Color comparison chart not included.
Specifications
- Description:pH Indicator Papers, Litmus Blue, Laboratory, Strips
- Grade:Laboratory
- Form:Strips



