Orders LinkContactUsLinkComponent
Ward's® Albite
Ward's® Albite
Storage box
Catalog # 470147-278
Supplier #:  196883
Ward's® Albite
Catalog # 470147-278
Ward's® Albite
Ward's® Albite
Storage box
Price
$1.50
Log in for availability
Each
Product Details & Documents
Feldspar group; white to gray, sodium plagioclase cleavages; some faces may show twinning.
  • Collected in Ontario, Canada
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25 to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Specifications
  • Description:
    Storage box
  • Cat. No.:
    470147-278