Slide Grid
Creatures, for NYS Test Kits (Centipede,Fly,Spider,Dragonfly,Bristletail,Ant)
Catalog # 470125-942
Supplier: Skywill Far East Industries
Supplier #: 2984200
Jump to:
Product Details & Documents
Microscopy tool to estimate size and counts.
- Microscopy study and research tool
- Improve your counting estimates
Use this slide grid as a tool to count or measure cells or organisms or to estimate the size of microscopic organisms. The grid is 1 cm square with mm divisions.
Specifications
- Description:Creatures, for NYS Test Kits (Centipede,Fly,Spider,Dragonfly,Bristletail,Ant)