Order Entry
United States
Orders LinkContactUsLinkComponent
Slide Grid

Slide Grid

Sorting Chart, 8 1/2 x 14" Laminated
Catalog # 470092-730
Supplier:   IMAGEN
Supplier #:  2945100
Slide Grid
Catalog # 470092-730
Supplier:  IMAGEN
Slide Grid

Sorting Chart, 8 1/2 x 14" Laminated

Clearance
$2.85
Sale

Valid until 31/12/2026

Log in for availability
Each
Product Details & Documents

Microscopy tool to estimate size and counts.

  • Microscopy study and research tool
  • Improve your counting estimates

Use this slide grid as a tool to count or measure cells or organisms or to estimate the size of microscopic organisms. The grid is 1 cm square with mm divisions.

Specifications
  • Description:
    Sorting Chart, 8 1/2 x 14" Laminated