Orders LinkContactUsLinkComponent
pH Indicator Papers
pH Indicator Papers
Catalog # 470151-048
pH Indicator Papers
Catalog # 470151-048
Supplier:  
pH Indicator Papers
pH Indicator Papers
Discontinued
Product Details & Documents
pH Test Papers in your Choice of Two Indicator Colors.
  • Easy to read
  • Vial of 100
Litmus red paper changes to blue at pH 8.1. Color comparison chart not included.