Orders LinkContactUsLinkComponent
Boreal Science Compound Beginner Microscopes
Boreal Science Compound Beginner Microscopes
Catalog # 470145-712
Boreal Science Compound Beginner Microscopes
Catalog # 470145-712
Supplier:  
Boreal Science Compound Beginner Microscopes
Boreal Science Compound Beginner Microscopes
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
MICROSCOPE EM PRIMARY CL COARSE Catalog #470014-506
Product 470014-506 is a replacement for 470145-712 but varies by Brand, Packaging or Quantity.
Product Details & Documents
Boreal beginner microscopes are ideal for introducing students to microscopy.
  • All-Glass-Coated Optics
  • All-Metal Construction
  • Parfocal and Parcentered
  • Objectives: 4x, 10x, and 40x
  • Boreal Lifetime Warranty
It's scaled down for younger students and is simple to use, but it also prepares students for the transition to high school instruction.
Rugged metal components are engineered for years of reliable performance. Student-proof features minimize loss of parts.
Ordering information: This product includes a manufacturer's warranty.