Botany Prepared Microscope Slides
Leaf - Lily, Cross Sectiontion, Saf and FG, Plant, Lilium sp.
Catalog # 470126-886
Supplier: C & A Scientific
Supplier #: 2H-192
Jump to:
Product Details & Documents
Great for beginners.
- Professionally stained samples for good visualization
- Perfect for your introductory biology classrooms
Stock-up on these affordable prepared microscope slides for your classroom. These slides are a great way to introduce your students to viewing samples under a microscope.
: These slides are made with care, by our manufacturing partners worldwide.
Specifications
- Type:Plant, Lilium sp.
- Description:Leaf - Lily
- Stain:Saf and FG
- Orientation:Cross Sectiontion