Order Entry
United States
Orders LinkContactUsLinkComponent
BactiSterilizer Microsterilizers

BactiSterilizer Microsterilizers

Slide Dryer (for 0.55" dia. units only)
Catalog # 470124-056
Supplier:   Ward's
Supplier #:  #B1000-03
BactiSterilizer Microsterilizers
Catalog # 470124-056
Supplier:  Ward's
BactiSterilizer Microsterilizers
BactiSterilizer Microsterilizers

Slide Dryer (for 0.55" dia. units only)

Price
$183.70
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Quick Benchtop Sterilization.

  • Complete sterilization in 5 to 7 seconds
  • Available in multiple sizes, with optional 'tilt' feature for easier use
  • Core enclosed in stainless steel guard chamber for safety
  • Ideal for platinum loops, needles and tube mouths
  • Maximum temperature 815 °C (1500 °F)

These line of infrared micro-sterilizers have asbestos-free core elements that use infrared heating to produce 815 °C (1500 °F). Choose between a standard-sized stationary barrel, or a tilt-able, adjustable-height unit available with a standard-size or XL barrel. For research use only.

Specifications
  • Description:
    Slide Dryer (for 0.55" dia. units only)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.