Orders LinkContactUsLinkComponent
Edvotek® Variable Micropipettes
Edvotek® Variable Micropipettes
Pipette Carousel Stand for Six Pipets, Light Gray
Catalog # 470121-052
Supplier:   Edvotek
Supplier #:  796
Edvotek® Variable Micropipettes
Catalog # 470121-052
Supplier:  Edvotek
Edvotek® Variable Micropipettes
Edvotek® Variable Micropipettes
Pipette Carousel Stand for Six Pipets, Light Gray
Price
$72.59
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Our newly re-designed variable micropipettes are sturdily designed with volumes ranging from 0.1 to 5000 µl.
  • New and improved
  • High quality, same price
  • More durable and ergonomic
  • Lifetime warranty
  • Light gray color
They are easy to use, highly accurate and use standard micropipette tips. The volume is easily selected by twisting the top. The lightweight design and tip ejector make operation fast and easy.
Ordering information: A tool and instructions are included for self-calibration.
Specifications
  • Description:
    Pipette Carousel Stand for Six Pipets, Light Gray