Orders LinkContactUsLinkComponent
pH Indicator Papers
pH Indicator Papers
Catalog # 470024-850
pH Indicator Papers
Catalog # 470024-850
Supplier:  
pH Indicator Papers
pH Indicator Papers
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PAPER PH INDICATOR PHEPH LN 4.8M 3ROLLS Catalog #EM1.09521.0003
Product EM1.09521.0003 is similar to 470024-850 but manufacturer and specifications may vary.
Product Details & Documents
pH Test Papers in your Choice of Two Indicator Colors.
  • Easy to read
  • Vial of 100
Litmus red paper changes to blue at pH 8.1. Color comparison chart not included.