Orders LinkContactUsLinkComponent
pH Indicator Papers
  470024-816
pH Indicator Papers
  470024-816
 :  
pH Indicator Papers
pH Indicator Papers
pH Test Papers in your Choice of Two Indicator Colors.
  • Easy to read
  • Vial of 100
Litmus red paper changes to blue at pH 8.1. Color comparison chart not included.