Orders LinkContactUsLinkComponent
Ward's® Ballistic Angle Set
Ward's® Ballistic Angle Set
WARD’S Economy Trajectory Rods
Catalog # 470020-132
Supplier:   Dynalon
Supplier #:  8551-3864
Ward's® Ballistic Angle Set
Catalog # 470020-132
Supplier:  Dynalon
Ward's® Ballistic Angle Set
Ward's® Ballistic Angle Set
WARD’S Economy Trajectory Rods
Clearance
$74.75
Log in for availability
Kit of 1
Product Details & Documents
Simulate Bullet Holes for Trajectory Determinations
Simulate Bullet Holes for Trajectory Determinations
Follow the path of a bullet from weapon to victim or surface using the components of this ballistics set. Two sets of predrilled plastic blocks simulate the holes produced by 9 mm and .45 caliber bullets entering at 30, 45, 60, and 90 degree angles. Mount the blocks on a wall or any surface and determine trajectory using either trajectory rods or string, available separately. Set of 8 blocks.
WARD’S Economy Trajectory Rods
These affordable polypropylene trajectory rods come with interchangeable tips designed to fit either a 9 mm or .45 caliber bullet hole. Size of each rod: 7 1/4"L x 1/4" dia. Set of four rods.
Ballistic Angle and Trajectory Rod Set
Buy Both the Ballistic Angle Set and Economy Trajectory Rods and Save
Specifications
  • Description:
    WARD’S Economy Trajectory Rods