Replica Diffraction Gratings
Diffraction grating slide, 5400 lines/cm, pack of 25
Catalog # 470006-576
Supplier #: 1503-00/25
Jump to:
Discontinued
Product Details & Documents
These simple slide-mounted diffraction gratings are great for experiments with lasers and LEDs.
Mounted in 2×2" slides
- 5360 lines per cm
These replica transmission diffraction gratings are transparent and irridescent. Approximately 5400 lines per centimeter, they are perfect for Young's double slit experiment or for viewing gas emission spectra. Mounted in unmarked cardboard slide holders, they are inexpensive and durable.
Specifications
- Description:Diffraction grating slide, 5400 lines/cm, pack of 25