Order Entry
United States
Orders LinkContactUsLinkComponent
Graph Papers

Graph Papers

Graph paper, metric coordinates, 22 cm x 28 cm
Catalog # 470005-678
Supplier:   IMAGEN
Supplier #:  6264405
Graph Papers
Catalog # 470005-678
Supplier:  IMAGEN
Graph Papers
Graph Papers

Graph paper, metric coordinates, 22 cm x 28 cm

Clearance
$11.15
Sale

Valid until 31/12/9999

Log in for availability
Pack of 100
Product Details & Documents

The logarithmic paper has a one cycle semi-log chart.

The metric graph paper is ruled 10 lines per cm with the fifth and tenth line accented. The polar coordinate paper has every degree indicated and every 10° accented. Each is three-hole punched and measures 22×28 cm. Packages of 100.

Specifications
  • Description:
    Graph paper, metric coordinates, 22 cm x 28 cm