Triangular Files
Triangular file, File: 4", 7"
Catalog # 470005-474
Supplier: United Scientific Supplies
Supplier #: TRF004
Jump to:
Product Details & Documents
Useful for metal filing or glass cutting.
- Wooden handle for easy gripping
- Hardened triple sides
Triangular file has a tapered shape, and is mounted on a hardwood handle for easy gripping. For general laboratory use, including to score glass before cutting.
Specifications
- Description:Triangular file
- Length:File: 4"
- Overall length:7"
Accessories
Product Information
- Catalog #LengthOverall lengthAvailabilityPrice
- Triangular file



