Pocket Laser Pointer
Pocket laser pointer, red
Catalog # 470003-492
Supplier: THE BEST CONNECTION INC
Supplier #: 4012
Jump to:
Product Details & Documents
Class IIIa red laser pointer for optics experiments.
- Classically designed laser pointer
- Range of over 500 yards
- Maximum power: 5 mW (Class IIIa)
- Approximate wavelength: 650 nm
- Made of aircraft aluminum
- Dimensions: 5³/₈×⁷/₁₆"
- Weight: 4.7 oz.
Ideal for indoor and outdoor use; for presentations and instruction.
Ordering information: Includes 2 AAA batteries and a plastic gift/storage box.
Specifications
- Description:Pocket laser pointer, red
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



