Order Entry
United States
Orders LinkContactUsLinkComponent
Dropper Assemblies

Dropper Assemblies

Assembly droppers, For 1 oz. bottles, Glass, 3×70 mm
Catalog # 470020-798
Supplier:   GSC International
Supplier #:  407-5-DZ
Dropper Assemblies
Catalog # 470020-798
Supplier:  GSC International
Dropper Assemblies
Dropper Assemblies

Assembly droppers, For 1 oz. bottles, Glass, 3×70 mm

Price
$16.90
Log in for availability
Pack of 12
Product Details & Documents

We sell many different sizes of dropper assemblies, varying by size, capacity, and the size of threaded neck they are designed to fit.

  • 24/400 neck size
  • Glass pipette with rubber bulb and plastic cap

Straight-talking and practical, our 24/400 neck size glass pipette with rubber bulb and plastic cap is the reliable dropper assembly you need in your lab. This no-frills design features a 20/400 cap that gets the job done without fuss.

The clear, flint glass construction is durable, while the simple rubber bulb and plastic cap provide a secure, leak-proof seal. Whether you're measuring out precious samples or refilling reagents, this dropper assembly puts functionality first. No flashy features, just a solid, trustworthy tool to keep your lab operations running smoothly. Get back to work with this honest, hardworking dropper - no extra bells and whistles required.

Specifications
  • Ø×L:
    3×70 mm
  • Description:
    Assembly droppers
  • Material:
    Glass
  • Size:
    For 1 oz. bottles
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.