Order Entry
United States
Orders LinkContactUsLinkComponent
BioCam Cell Structure And Function Charts

BioCam Cell Structure And Function Charts

Meiosis Chart, Laminated, 61 cm, 45.7 cm
Catalog # 470008-124
Supplier #:  WC09
BioCam Cell Structure And Function Charts
Catalog # 470008-124
BioCam Cell Structure And Function Charts
BioCam Cell Structure And Function Charts

Meiosis Chart, Laminated, 61 cm, 45.7 cm

Price
$35.50
Log in for availability
Each
Product Details & Documents

A mixture of photo-micrographs, detailed artwork and diagrams make these charts excellent for supporting your unit on cells and cell division.

  • Highly detailed

Explore cells with these charts.

Specifications
  • Width:
    45.7 cm
  • Description:
    Meiosis Chart
  • Length:
    61 cm
  • Finish:
    Laminated
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.